Learn More
CPC Scientific Ac-Ile-Glu-Thr-Asp-CHO 1MG
SDP
Supplier:  CPC Scientific CASP057A
Encompass
Peptide aldehyde that corresponds to one of the cleavage sites of the inactive 32 kD caspase-3 precursor (amino acids 172-175). In a concentration range of 0.5 ?M, it blocks the formation of the p17 subunit and concomitantly induces the accumulation of the 32kD precursor.SEQUENCE: Ac-Ile-Glu-Thr-Asp-CHOONE-LETTER SEQUENCE: Ac-IETD-CHOMOLECULAR FORMULA: C21H34N4O10MOLECULAR WEIGHT: 502.5STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [191338-86-0]SYNONYMS: RESEARCH AREA: ApoptosisREFERENCES:Z. Han et al, J. Biol Chem., 272, 13422 (1997)M.Garcia-Calvo et al., J. Biol. Chem., 273, 32608 (1998)SKU(s): CASP-057A, CASP-057B
Catalog No. 50-210-9020