Learn More
Enzo Life Sciences Ac-IETD-CHO (5mg). CAS: 191338-86-0
SDP
Catalog No. 502008254
Encompass_Preferred
Caspase and granzyme inhibitor. Reversible inhibitor of caspase-6, -8 and granzyme B. Also inhibits caspase-10. Alternative name: Caspase 8 inhibitor (aldehyde), Granzyme B inhibitor (aldehyde). Formula: C21H34N4O10. MW: 502.5. Purity: ≥96% (HPLC). Formulation: Lyophilized. Peptide content: 70-90%. Solubility: Soluble in distilled water at 10 mg/mL and in DMSO at 1 mg/mL. Long Term Storage: -20°C. Use/Stability: Use always fresh solutions.
Catalog No. 50-200-8254 Supplier Enzo Life Sciences Supplier No. ALX260043M005
Caspase and granzyme inhibitor. Reversible inhibitor of caspase-6, -8 and granzyme B. Also inhibits caspase-10. Alternative name: Caspase 8 inhibitor (aldehyde), Granzyme B inhibitor (aldehyde). Formula: C21H34N4O10. MW: 502.5. Purity: ≥96% (HPLC). Formulation: Lyophilized. Peptide content: 70-90%. Solubility: Soluble in distilled water at 10 mg/mL and in DMSO at 1 mg/mL. Long Term Storage: -20°C. Use/Stability: Use always fresh solutions.