Learn More
Medchemexpress LLC Ac-IHIHIQI-NH2 | 1582727-85-2 | 99.6% | 914.11 | 1 MG
SDP
Catalog No. 5000398586
Encompass_Preferred
Ac-IHIHIQI-NH2 is a fibril-forming heptapeptide with high catalytic activity to laccase mimics. It exhibits selectivity for hydrophobic p-nitrophenyl (ONp) ester substrates in the process of self-assembly.
  • Fibril-forming heptapeptide
  • High catalytic activity to laccase mimics
  • Exhibits selectivity for hydrophobic p-nitrophenyl (ONp) ester substrates in the process of self-assembly
  • Catalyzes p-nitrophenyl acetate (pNPA) hydrolysis in the presence of Zn2+ with a kcat/KM of 62 M-1s-1 at pH 8 (In Vitro)
Catalog No. 50-003-98586 Supplier Medchemexpress LLC Supplier No. HYP41471MG
Ac-IHIHIQI-NH2 is a fibril-forming heptapeptide with high catalytic activity to laccase mimics. It exhibits selectivity for hydrophobic p-nitrophenyl (ONp) ester substrates in the process of self-assembly.
  • Fibril-forming heptapeptide
  • High catalytic activity to laccase mimics
  • Exhibits selectivity for hydrophobic p-nitrophenyl (ONp) ester substrates in the process of self-assembly
  • Catalyzes p-nitrophenyl acetate (pNPA) hydrolysis in the presence of Zn2+ with a kcat/KM of 62 M-1s-1 at pH 8 (In Vitro)