Learn More
CPC Scientific Ac-Ile-Glu-Pro-Asp-pNA 25MG
SDP
Supplier:  CPC Scientific CASP078C
Encompass
Caspase substrate that contains the granzyme B cleavage site. It is additionally proposed as a potential granzyme B target and the optimal P4-P1 substrate sequence for granzyme B.ONE-LETTER SEQUENCE: Ac-IEPD-pNAMOLECULAR FORMULA: C28H38N6O11MOLECULAR WEIGHT:634.7STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [216757-29-8], SYNONYMS: Ac-IEPD-pNARESEARCH AREA: ApoptosisREFERENCES: J.L. Harris et al., J. Biol. Chem., 273, 27364 (1998)
Catalog No. 50-210-9066