Learn More
Ubpbio A fluorogenic substrate of the trypsin-like activity of proteasomes
Catalog No. 502339365
Encompass_Preferred
Ac-KQL-AMC is a fluorogenic substrate of the trypsin-like activity of both constitutive and immuno proteasomes. The AMC fluorescence can be detected by a fluorimeter or a plate reader using excitation/emission wavelengths at 360 nm/460 nm, respectively. This substrate can be used to determine the trypsin-like activity of proteasomes. Working concentration is 50 - 200 µM.
Catalog No. 50-233-9365 Supplier Ubpbio Supplier No. G3110
Ac-KQL-AMC is a fluorogenic substrate of the trypsin-like activity of both constitutive and immuno proteasomes. The AMC fluorescence can be detected by a fluorimeter or a plate reader using excitation/emission wavelengths at 360 nm/460 nm, respectively. This substrate can be used to determine the trypsin-like activity of proteasomes. Working concentration is 50 - 200 µM.