Learn More
Enzo Life Sciences Ac-LEHD-AFC (10mg). CAS: 210345-03-2
SDP
Catalog No. 502008327
Encompass_Preferred
Fluorogenic substrate for caspase-9. Alternative name: Caspase 9 substrate (fluorogenic). Formula: C33H38F3N7O11. MW: 765.7. Purity: ≥96% (HPLC). Formulation: Lyophilized. Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8327 Supplier Enzo Life Sciences Supplier No. ALX260116M010
Fluorogenic substrate for caspase-9. Alternative name: Caspase 9 substrate (fluorogenic). Formula: C33H38F3N7O11. MW: 765.7. Purity: ≥96% (HPLC). Formulation: Lyophilized. Solubility: Soluble in DMSO. Long Term Storage: -20°C.