Learn More
Enzo Life Sciences Ac-LEHD-AFC (5mg). CAS: 210345-03-2
SDP
Supplier:  Enzo Life Sciences ALX260116M005
Encompass_Preferred
Caspase-9 substrate. Fluorogenic substrate for caspase-9. Alternative name: Caspase 9 substrate (fluorogenic). Formula: C33H38F3N7O11. MW: 765.7. Purity: ≥96% (HPLC). Formulation: Lyophilized. Solubility: Soluble in DMSO. Long Term Storage: -20°C. Handling: Protect from light.
Catalog No. 50-200-8326