Learn More
Enzo Life Sciences Ac-LEHD-AMC (5mg). CAS: 292633-16-0
SDP
Catalog No. 502008297
Encompass_Preferred
Caspase-9 substrate. Fluorogenic substrate for caspase-9 and related cysteine proteases. The LEHD sequence is derived from combinatorial experiments. Ex.:340-360nm, Em.:440-460nm. Alternative name: Caspase 9 substrate (fluorogenic). Formula: C33H41N7O11. MW: 711.7. Purity: ≥98% HPLC. Appearance: White to off-white powder. Peptide content: 75-95%. Solubility: Soluble in water,  DMSO or phosphate buffer, pH 7.5. Long Term Storage: -20°C. Handling: Keep cool and dry.
Catalog No. 50-200-8297 Supplier Enzo Life Sciences Supplier No. ALX260080M005
Add to Cart
Add to Cart
Caspase-9 substrate. Fluorogenic substrate for caspase-9 and related cysteine proteases. The LEHD sequence is derived from combinatorial experiments. Ex.:340-360nm, Em.:440-460nm. Alternative name: Caspase 9 substrate (fluorogenic). Formula: C33H41N7O11. MW: 711.7. Purity: ≥98% HPLC. Appearance: White to off-white powder. Peptide content: 75-95%. Solubility: Soluble in water,  DMSO or phosphate buffer, pH 7.5. Long Term Storage: -20°C. Handling: Keep cool and dry.