Learn More
CPC Scientific Ac-Leu-Glu-His-Asp-CHO 5MG
SDP
Catalog No. 502109075
Encompass
ONE-LETTER SEQUENCE: Ac-LEHD-CHOMOLECULAR FORMULA: C23H34N6O9MOLECULAR WEIGHT:538.56STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [319494-38-7], SYNONYMS: Ac-Leu-Glu-His-Asp-aldehyde (pseudo acid)RESEARCH AREA: Apoptosis
Catalog No. 50-210-9075 Supplier CPC Scientific Supplier No. CASP082B
Add to Cart
Add to Cart
ONE-LETTER SEQUENCE: Ac-LEHD-CHOMOLECULAR FORMULA: C23H34N6O9MOLECULAR WEIGHT:538.56STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [319494-38-7], SYNONYMS: Ac-Leu-Glu-His-Asp-aldehyde (pseudo acid)RESEARCH AREA: Apoptosis