Learn More
Enzo Life Sciences Ac-LEHD-CHO (5mg)
SDP
Supplier:  Enzo Life Sciences ALX260079M005
Encompass_Preferred
Caspase-9 inhibitor. Reversible aldehyde inhibitor of caspase-9 and related cysteine proteases. Sequence is from combinatorial experiments. Alternative name: Caspase 9 inhibitor (aldehyde). Formula: C23H34N6O9. MW: 538.6. Purity: ≥95%. Peptide content: 75-95%. Solubility: Soluble in water or DMSO. Further dilute with buffer, pH 7.5. Long Term Storage: -20°C. Use/Stability: Keep sealed after removing from the freezer until its temperature equilibrates with room temperature. Dissolve only amount needed at a time to avoid decomposition in solution. Handling: Keep cool and dry.
Catalog No. 50-200-8295