Learn More
Enzo Life Sciences Ac-LEHD-pNA (5mg)
SDP
Supplier:  Enzo Life Sciences ALX260081M005
Encompass_Preferred
Caspase-9 substrate. Chromogenic substrate for caspase-9 and cysteine proteases. Similar to Ac-LEHD-AMC (ALX-260-080) and Ac-LEHD-AFC (ALX-260-116) but cleavage is monitored colorimetrically by absorbance at 405nm. Alternative name: Caspase 9 substrate (chromogenic). Formula: C29H38N8O11. MW: 674.7. Purity: ≥99% (HPLC). Formulation: Lyophilized. Peptide content: 75-95%. Solubility: Soluble in water or  DMSO. Dilute with buffer, pH 7.5. Long Term Storage: -20°C.
Catalog No. 50-200-8299