Learn More
Medchemexpress LLC Ac-LEHD-pNA | 921771-40-6 | 99.6% | 674.66 | 10 MG
SDP
Catalog No. 5000396954
Encompass_Preferred
Ac-LEHD-pNA is a biologically active peptide that serves as a Caspase-9 substrate. This pNA (4-nitroaniline)-derived caspase substrate is widely utilized for the colorimetric detection of various caspase activities. The cleavage of pNA peptides by caspases generates pNA, which is monitored colorimetrically at approximately 405 nm.
  • Serves as a Caspase-9 substrate
  • Used for colorimetric detection of caspase activities
  • Generates pNA upon cleavage for monitoring at ~405 nm
Catalog No. 50-003-96954 Supplier Medchemexpress LLC Supplier No. HYP534810MG
Ac-LEHD-pNA is a biologically active peptide that serves as a Caspase-9 substrate. This pNA (4-nitroaniline)-derived caspase substrate is widely utilized for the colorimetric detection of various caspase activities. The cleavage of pNA peptides by caspases generates pNA, which is monitored colorimetrically at approximately 405 nm.
  • Serves as a Caspase-9 substrate
  • Used for colorimetric detection of caspase activities
  • Generates pNA upon cleavage for monitoring at ~405 nm