Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-LEHD-pNA | 921771-40-6 | 99.6% | 674.66 | 25 MG
SDP

Catalog No. 5000397419
Encompass_Preferred

Ac-LEHD-pNA is a biologically active peptide and a Caspase-9 substrate. pNA (4-nitroaniline)-derived caspase substrates are widely used for the colorimetric detection of various caspase activities. The cleavage of pNA peptides by caspases generates pNA, which is monitored colorimetrically at approximately 405 nm, with a maximum absorption around 408 nm.

  • Caspase-9 substrate
  • Used for colorimetric detection of caspase activities
  • Generates pNA upon cleavage
  • Monitored colorimetrically at ~405 nm
  • Maximum absorption at ~408 nm

Catalog No. 50-003-97419 Supplier Medchemexpress LLC Supplier No. 25MGHYP5348
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ac-LEHD-pNA is a biologically active peptide and a Caspase-9 substrate. pNA (4-nitroaniline)-derived caspase substrates are widely used for the colorimetric detection of various caspase activities. The cleavage of pNA peptides by caspases generates pNA, which is monitored colorimetrically at approximately 405 nm, with a maximum absorption around 408 nm.

  • Caspase-9 substrate
  • Used for colorimetric detection of caspase activities
  • Generates pNA upon cleavage
  • Monitored colorimetrically at ~405 nm
  • Maximum absorption at ~408 nm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.