Learn More
Medchemexpress LLC Ac-LEHD-pNA | 921771-40-6 | 99.6% | 674.66 | 5 MG
SDP
Catalog No. 5000398932
Encompass_Preferred
Ac-LEHD-pNA is a biologically active peptide that serves as a Caspase-9 substrate. pNA (4-nitroaniline)-derived caspase substrates are commonly used for the colorimetric detection of various caspase activities. The cleavage of pNA peptides by caspases generates pNA, which can be monitored colorimetrically at approximately 405 nm. pNA has a maximum absorption around 408 nm.
  • Biological active peptide
  • Caspase-9 substrate
  • Used for colorimetric detection of caspase activities
Catalog No. 50-003-98932 Supplier Medchemexpress LLC Supplier No. HYP53485MG
Ac-LEHD-pNA is a biologically active peptide that serves as a Caspase-9 substrate. pNA (4-nitroaniline)-derived caspase substrates are commonly used for the colorimetric detection of various caspase activities. The cleavage of pNA peptides by caspases generates pNA, which can be monitored colorimetrically at approximately 405 nm. pNA has a maximum absorption around 408 nm.
  • Biological active peptide
  • Caspase-9 substrate
  • Used for colorimetric detection of caspase activities