Learn More
Cayman Chemical Ac-LETD-AFC 10mg
SDP
Catalog No. 502972308
Encompass_Preferred
A fluorogenic substrate that can be cleaved specifically by caspase-8
Catalog No. 50-297-2308 Supplier Cayman Chemical Supplier No. 1499110 MG
A fluorogenic substrate that can be cleaved specifically by caspase-8