Learn More
Cayman Chemical Ac-LETD-AFC 1mg
SDP
Catalog No. 502970023
Encompass_Preferred
A fluorogenic substrate that can be cleaved specifically by caspase-8
Catalog No. 50-297-0023 Supplier Cayman Chemical Supplier No. 149911 MG
A fluorogenic substrate that can be cleaved specifically by caspase-8