Learn More
Cayman Chemical Ac-LETD-AFC 5mg
SDP
Catalog No. 502970024
Encompass_Preferred
A fluorogenic substrate that can be cleaved specifically by caspase-8
Catalog No. 50-297-0024 Supplier Cayman Chemical Supplier No. 149915 MG
A fluorogenic substrate that can be cleaved specifically by caspase-8