Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-Leu-Arg-AMC | 929621-79-4 | 98.2% | 486.56 | 25 MG
SDP

Catalog No. 5000426978
Encompass_Preferred

Ac-Leu-Arg-AMC is a fluorogenic peptide substrate used in research. It has specific excitation and emission wavelengths, making it suitable for fluorescent assays.

  • Fluorogenic peptide substrate
  • Excitation (Ex): 390
  • Emission (Em): 480
  • Soluble in DMSO (300 mg/mL)
  • Store powder sealed, away from moisture and light for extended periods
  • Store in solvent as aliquots at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-26978 Supplier Medchemexpress LLC Supplier No. 25MGHYP1448
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ac-Leu-Arg-AMC is a fluorogenic peptide substrate used in research. It has specific excitation and emission wavelengths, making it suitable for fluorescent assays.

  • Fluorogenic peptide substrate
  • Excitation (Ex): 390
  • Emission (Em): 480
  • Soluble in DMSO (300 mg/mL)
  • Store powder sealed, away from moisture and light for extended periods
  • Store in solvent as aliquots at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.