Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-Leu-Arg-AMC | 929621-79-4 | 98.2% | 486.56 | 5 MG
SDP

Catalog No. 5000422114
Encompass_Preferred

Ac-Leu-Arg-AMC is a fluorogenic peptide substrate used in various research applications. It is supplied as a solid with a purity of 98.2% and a molecular weight of 486.56. This compound is known for its fluorescent properties with an excitation wavelength of 390 nm and emission at 480 nm, making it suitable for fluorescence-based assays.

  • Fluorogenic peptide substrate
  • White to off-white solid appearance
  • Excitation (Ex) at 390 nm
  • Emission (Em) at 480 nm
  • Soluble in DMSO (300 mg/mL)

Catalog No. 50-004-22114 Supplier Medchemexpress LLC Supplier No. HYP14485MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ac-Leu-Arg-AMC is a fluorogenic peptide substrate used in various research applications. It is supplied as a solid with a purity of 98.2% and a molecular weight of 486.56. This compound is known for its fluorescent properties with an excitation wavelength of 390 nm and emission at 480 nm, making it suitable for fluorescence-based assays.

  • Fluorogenic peptide substrate
  • White to off-white solid appearance
  • Excitation (Ex) at 390 nm
  • Emission (Em) at 480 nm
  • Soluble in DMSO (300 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.