Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-Leu-Glu-Val-Asp-CHO | 402832-01-3 | 98.4% | C22H36N4O9 | 10MG
SDP

Catalog No. 5000259362
Encompass_Preferred

Ac-LEVD-CHO is a peptide aldehyde inhibitor of caspase-4 intended for research use. It has the sequence Ac-Leu-Glu-Val-Asp-CHO and is supplied with high purity for biochemical and cell-based assays.

  • Peptide aldehyde inhibitor of caspase-4.
  • Sequence: Ac-Leu-Glu-Val-Asp-CHO for specific substrate recognition.
  • High purity (98.4%) suitable for biochemical studies.
  • Molecular weight 500.54 g·mol⁻¹ and formula C22H36N4O9.
  • Available in small research quantities (1-100 mg) to support assay development.

Catalog No. 50-002-59362 Supplier Medchemexpress LLC Supplier No. HY13672710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ac-LEVD-CHO is a peptide aldehyde inhibitor of caspase-4 intended for research use. It has the sequence Ac-Leu-Glu-Val-Asp-CHO and is supplied with high purity for biochemical and cell-based assays.

  • Peptide aldehyde inhibitor of caspase-4.
  • Sequence: Ac-Leu-Glu-Val-Asp-CHO for specific substrate recognition.
  • High purity (98.4%) suitable for biochemical studies.
  • Molecular weight 500.54 g·mol⁻¹ and formula C22H36N4O9.
  • Available in small research quantities (1-100 mg) to support assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.