Learn More
CPC Scientific Ac-Leu-Glu-Val-Asp-CHO 5MG
SDP
Supplier:  CPC Scientific CASP067B
Encompass
Inhibitor of caspase-4 (ICH-2).ONE-LETTER SEQUENCE: Ac-LEVD-CHOMOLECULAR FORMULA: C22H36N4O9MOLECULAR WEIGHT:500.55STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [402832-01-3], RESEARCH AREA: ApoptosisREFERENCES: R.V. Talanian et al., J. Biol. Chem. , 272, 9677 (1997)
Catalog No. 50-210-9042