Learn More
Enzo Life Sciences Ac-Leu-Leu-Met-al (5mg). CAS: 110115-07-6
SDP
Supplier:  Enzo Life Sciences BMLPI1000005
Encompass_Preferred
Cell-permeable, peptide aldehyde inhibitor of calpain and other neutral cysteine proteases. Potent inhibitor of cathepsin L (Ki=0.6nM) and cathepsin B (Ki=100nM). Alternative name: ALLM, Ac-LLM-CHO, Calpain inhibitor 2. Purity: ≥95% (HPLC). Solubility: 5mg/ml soluble in 100% ethanol, methanol or DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-1435