Learn More
Enzo Life Sciences Ac-Leu-Leu-Nle-al (25mg). CAS: 110044-82-1
SDP
Supplier:  Enzo Life Sciences BMLP1200025
Encompass_Preferred
Cell-permeable, peptide aldehyde inhibitor of calpain I (Ki=190nM), calpain II (Ki=150nM), cathepsin L (Ki=0.5nM) and other neutral cysteine proteases. Inhibits cell cycle progression at G1/S and metaphase/anaphase in CHO cells by inhibiting cyclin B degradation. Also stimulates HMG-CoA synthase transcription by inhibiting degradation of active SREBP-1 (sterol regulatory element-binding protein 1). Protects against neuronal damage caused by hypoxia and ischemia. Inhibits apoptosis in thymocytes and metamyelocytes. Prevents nitric oxide production by activated macrophages by interfering with the transcription of inducible nitric oxide synthase . Inhibits proteolytic degradation of IκBα and IκBß in RAW macrophages induced with LPS. It also prolong association of MHC class I molecules with the transporters associated with antigen processing. Alternative name: ALLN, Calpain inhibitor I, Ac-LL-norleucinal. Purity: ≥95% (HPLC). Solubility: Soluble in ethanol (5 mg/ml) or DMSO (10 mg/ml).
Catalog No. 50-201-1236