Learn More
Carbosynth LLC. Ac-Leu-Leu-Nle-H (aldehyde), 5mg, MW: (M.W. 383.54), Molecular formula: C20H37N3O4, CAS# [110044-82-1]
SDP
Supplier:  Carbosynth LLC. IAL3671PI5MG
Encompass_Preferred
This item has a minimum order quantity of 3 per supplier requirements
Synonyms: ALLN, MG 101 Acetyl-L-Leucyl-L-Leucyl-L-Norleucinal, Application: Inhibitor for Calpain I and Proteasome, Storage Temperature: -20C, References: T. Sasaki, M. Kishi, M. Saito, T. Tanaka, N. Higuchi, E. Kominami, N. Katunuma, and T. Murachi, J. Enzyme Inhib., 3, 195 (1990).
Catalog No. 50-168-5807