Learn More
MP Biomedicals™ Ac-LEU-LEU-TYR-7-Amino-4-Trifluoromethylcoumarin
Shop All MP Biomedicals, Inc ProductsDescription
The AFC substrate is both chromogenic and fluorogenic. The specificity and sensitivity of AFC allows for the detection of enzyme activities in cell lysate and electrophoretic gels as the emission spectra of free and the conjugated AFC do not overlap. Excitation 400nm; Emission 505nm
Specifications
Specifications
| Color | White |
| Form | Solid |
| For Use With (Application) | Cell Migration Analysis, Cell Signaling Analysis |
| Quantity | 10mg |
| Molecular Weight (g/mol) | 660 |
| Storage Requirements | Keep cool and dry |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.