Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Ac-LEU-LEU-TYR-7-Amino-4-Trifluoromethylcoumarin

Catalog No. MP3AFC11625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP3AFC11625 25mg
MP3AFC11610 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP3AFC11625 Supplier MP Biomedicals™ Supplier No. 03AFC11625
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A fluorogenic substrate for Calpain

The AFC substrate is both chromogenic and fluorogenic. The specificity and sensitivity of AFC allows for the detection of enzyme activities in cell lysate and electrophoretic gels as the emission spectra of free and the conjugated AFC do not overlap. Excitation 400nm; Emission 505nm

TRUSTED_SUSTAINABILITY

Specifications

Color White
Form Solid
For Use With (Application) Cell Migration Analysis, Cell Signaling Analysis
Quantity 25mg
Molecular Weight (g/mol) 660
Storage Requirements Keep cool and dry
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.