Learn More
Bachem Ac-Leu-NH2
SDP
Supplier:  Bachem 4000614.0025
Encompass_Preferred
Ac-Leu-NH2, 28529-34-2, C8H16N2O2, Mr 172.23, 25g. The N-acetylamino acid amides NALA, NALMA, NAKA, NAQA, and NATA have been used as model compounds of varying polarity in hydration studies.
Catalog No. 50-258-949