Learn More
CPC Scientific Ac-Leu-Pro-Phe-Phe-Asp-NH2 1MG
SDP
Catalog No. 502108692
Encompass
beta-sheet breaker peptide iAbeta5p that crosses the blood-brain barrier at a higher rate than most proteins and peptides. It is known to be selectively taken up by the brain, thereby suggesting that the peptide is being specifically transported to the brain. A significant increase in neuronal survival and decrease in brain inflammation associated with the reduction of amyloid plaques in two different transgenic Alzheimer's disease (AD) models is additionally reported for this pentapeptide. Abeta (1-42) Fibrillogenesis Inhibitor.SEQUENCE: Ac-Leu-Pro-Phe-Phe-Asp-NH2ONE-LETTER SEQUENCE: Ac-LPFFD-NH2MOLECULAR FORMULA: C35H46N6O8MOLECULAR WEIGHT: 678.79STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [339990-02-2]SYNONYMS: Acetyl-(Pro,Asp)-Amyloid -Protein (17-21) amideRESEARCH AREA: Alzheimer's DiseaseREFERENCES:SKU(s): AMYD-033A, AMYD-033B
Catalog No. 50-210-8692 Supplier CPC Scientific Supplier No. AMYD033B
Add to Cart
Add to Cart
beta-sheet breaker peptide iAbeta5p that crosses the blood-brain barrier at a higher rate than most proteins and peptides. It is known to be selectively taken up by the brain, thereby suggesting that the peptide is being specifically transported to the brain. A significant increase in neuronal survival and decrease in brain inflammation associated with the reduction of amyloid plaques in two different transgenic Alzheimer's disease (AD) models is additionally reported for this pentapeptide. Abeta (1-42) Fibrillogenesis Inhibitor.SEQUENCE: Ac-Leu-Pro-Phe-Phe-Asp-NH2ONE-LETTER SEQUENCE: Ac-LPFFD-NH2MOLECULAR FORMULA: C35H46N6O8MOLECULAR WEIGHT: 678.79STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [339990-02-2]SYNONYMS: Acetyl-(Pro,Asp)-Amyloid -Protein (17-21) amideRESEARCH AREA: Alzheimer's DiseaseREFERENCES:SKU(s): AMYD-033A, AMYD-033B