Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-LEVD-AFC | 1092077-23-0 | 98.1% | 727.68 | 1 MG
SDP

Catalog No. 5000396628
Encompass_Preferred

Ac-LEVD-AFC is a capspase-4 fluorogenic substrate peptide for caspase-4 activity studies. This compound is for research use only.

  • Appears as a white to off-white solid
  • Molecular formula: C32H40F3N5O11
  • Sequence: Ac-Leu-Glu-Val-Asp-{AFC}
  • Store powder at -80°C for 2 years or -20°C for 1 year
  • Store in solvent at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture)

Catalog No. 50-003-96628 Supplier Medchemexpress LLC Supplier No. HYP31151MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ac-LEVD-AFC is a capspase-4 fluorogenic substrate peptide for caspase-4 activity studies. This compound is for research use only.

  • Appears as a white to off-white solid
  • Molecular formula: C32H40F3N5O11
  • Sequence: Ac-Leu-Glu-Val-Asp-{AFC}
  • Store powder at -80°C for 2 years or -20°C for 1 year
  • Store in solvent at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.