Learn More
Enzo Life Sciences Ac-LEVD-AFC (1mg)
SDP
Catalog No. 502008303
Encompass_Preferred
Caspase-4 substrate. Fluorogenic substrate for caspase-4. Similar to the fluorogenic substrate Ac-LEVD-AMC (ALX-260-083); the AFC fluorophore is more sensitive. Alternative name: Caspase 4 substrate (fluorogenic). Formula: C32H40F3N5O11. MW: 727.7. Purity: ≥98% (HPLC). Peptide content: 75-95%. Solubility: Soluble in DMSO; dilute with distilled water or buffer; also soluble in phosphate buffer, pH 7.5-8.0. Long Term Storage: -20°C. Use/Stability: Dissolve only as much as needed at one time because of the possibility of decomposition in solution. Store solution at -20°C. Protect from condensation: when taking the sample out of the container, wait until it warms up to room temperature, then open it. Handling: Keep cool and dry.
Catalog No. 50-200-8303 Supplier Enzo Life Sciences Supplier No. ALX260084M001
Caspase-4 substrate. Fluorogenic substrate for caspase-4. Similar to the fluorogenic substrate Ac-LEVD-AMC (ALX-260-083); the AFC fluorophore is more sensitive. Alternative name: Caspase 4 substrate (fluorogenic). Formula: C32H40F3N5O11. MW: 727.7. Purity: ≥98% (HPLC). Peptide content: 75-95%. Solubility: Soluble in DMSO; dilute with distilled water or buffer; also soluble in phosphate buffer, pH 7.5-8.0. Long Term Storage: -20°C. Use/Stability: Dissolve only as much as needed at one time because of the possibility of decomposition in solution. Store solution at -20°C. Protect from condensation: when taking the sample out of the container, wait until it warms up to room temperature, then open it. Handling: Keep cool and dry.