Learn More
Cayman Chemical Ac-LEVD-AFC, Purity: ≥95%, CAS: 1092077-23-0, 1 mg
SDP
Catalog No. 502937204
Encompass_Preferred
A fluorogenic substrate for caspase-4; AFC is released upon enzymatic cleavage by caspase-4 and its fluorescence can be used to quantify caspase-4 activity; ex/em = 400/505 nm, respectively
Catalog No. 50-293-7204 Supplier Cayman Chemical Supplier No. 288091 MG
A fluorogenic substrate for caspase-4; AFC is released upon enzymatic cleavage by caspase-4 and its fluorescence can be used to quantify caspase-4 activity; ex/em = 400/505 nm, respectively