Learn More
Enzo Life Sciences Ac-LEVD-AFC (5mg)
SDP
Supplier:  Enzo Life Sciences ALX260084M005
Encompass_Preferred
Fluorogenic substrate for caspase-4. Similar to the fluorogenic substrate Ac-LEVD-AMC (ALX-260-083); the AFC fluorophore is more sensitive. Alternative name: Caspase 4 substrate (fluorogenic). Formula: C32H40F3N5O11. MW: 727.7. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO; dilute with distilled water or buffer; also soluble in phosphate buffer, pH 7.5-8.0. Long Term Storage: -20°C.
Catalog No. 50-200-8304