Learn More
Cayman Chemical Ac-LEVD-AFC 5mg
SDP
Catalog No. 502939018
Encompass_Preferred
A fluorogenic substrate for caspase-4; AFC is released upon enzymatic cleavage by caspase-4 and its fluorescence can be used to quantify caspase-4 activity; ex/em = 400/505 nm, respectively
Catalog No. 50-293-9018 Supplier Cayman Chemical Supplier No. 288095 MG
A fluorogenic substrate for caspase-4; AFC is released upon enzymatic cleavage by caspase-4 and its fluorescence can be used to quantify caspase-4 activity; ex/em = 400/505 nm, respectively