Learn More
Enzo Life Sciences Ac-LEVD-AMC (5mg)
SDP
Supplier:  Enzo Life Sciences ALX260083M005
Encompass_Preferred
Fluorogenic substrate for caspase-4. Alternative name: Caspase 4 substrate (fluorogenic). Formula: C32H43N5O11. MW: 673.7. Purity: ≥99% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in DMSO. Dilute with distilled water or buffer, pH 7.5. Long Term Storage: -20°C. Handling: Keep cool and dry.
Catalog No. 50-200-8302