Learn More
Medchemexpress LLC Ac-LEVD-CHO | 402832-01-3 | 98.4% | 500.54 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1367271MG
Encompass_Preferred
Ac-LEVD-CHO is a caspase-4 inhibitor and a peptide with the sequence of Ac-Leu-Glu-Val-Asp-al. This product is provided as a white to off-white solid.
  • Inhibits caspase-4 activity
  • Peptide sequence: Ac-Leu-Glu-Val-Asp-al
  • Suitable for in vitro and in vivo studies
  • Available in various quantities for research needs
Catalog No. 50-003-48093