Learn More
Enzo Life Sciences Ac-LEVD-pNA (5mg)
SDP
Catalog No. 502008276
Encompass_Preferred
Chromogenic substrate for caspase-4. Alternative name: Caspase 4 substrate (chromogenic). Formula: C28H40N6O11. MW: 636.7. Purity: ≥99% (HPLC). Solubility: Soluble in buffer, pH 7.0-8.0 (e. g. TRIS, HEPES). Long Term Storage: -20°C.
Catalog No. 50-200-8276 Supplier Enzo Life Sciences Supplier No. ALX260061M005
Chromogenic substrate for caspase-4. Alternative name: Caspase 4 substrate (chromogenic). Formula: C28H40N6O11. MW: 636.7. Purity: ≥99% (HPLC). Solubility: Soluble in buffer, pH 7.0-8.0 (e. g. TRIS, HEPES). Long Term Storage: -20°C.