Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Ac-Lys-Pro-Arg-7-Amino-4-Trifluoromethylcoumarin

Catalog No. MP3AFC13025 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP3AFC13025 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP3AFC13025 Supplier MP Biomedicals™ Supplier No. 03AFC13025

missing translation for 'validMainframeMsg'

Fluorescent substrate for Tryptase

  • When hydrolyzed, free AFC produced in reaction can be quantified by by spectrophotometric detection or fluorometric detection
  • Acts as substrate for assay of Tryptase
TRUSTED_SUSTAINABILITY

Specifications

Color White
Form Solid
For Use With (Application) Substrate for Assay of Tryptase
Quantity 1 each
Product Type Ac-Lys-Pro-Arg-7-Amino-4-Trifluoromethylcoumarin
Storage Requirements Keep cool and dry, Desiccate AFC- or AMC130 in solid form at room temperature. Store DMSO/DMF solutions at -20°C. Material stable for at least one year, if stored as recommended.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.