Learn More
Carbosynth LLC. Ac-Lys-Val-Pro-Leu-ACC (5 mg vial), MF= C35H51N7O8
SDP
Supplier:  Carbosynth LLC. SGM3240V5MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Acetyl-L-Lysyl-L-Valyl-L-Prolyl-L-Leucine-4-Carbamoylmethylcoumaryl-7-Amide, Application: Selective Fluorogenic Substrate for Granzyme M, salt: Trifluoroacatate Salt, one letter code: Ac-KVPL-ACC , MW: 697.8
Catalog No. 50-195-2433