Learn More
CPC Scientific Ac-(NMe)Tyr-Val-Ala-Asp-CHO 1MG
SDP
Catalog No. 502108932
Encompass
ONE-LETTER SEQUENCE: Ac-(NMe)Tyr-Val-Ala-Asp-CHOMOLECULAR FORMULA: C24H34N4O8MOLECULAR WEIGHT:506.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [160806-26-8], RESEARCH AREA: ApoptosisREFERENCES: M.D.Mullican et al., Bioorg. Med. Chem. Letters, 4, 2359 (1994)
Catalog No. 50-210-8932 Supplier CPC Scientific Supplier No. CASP002B
Add to Cart
Add to Cart
ONE-LETTER SEQUENCE: Ac-(NMe)Tyr-Val-Ala-Asp-CHOMOLECULAR FORMULA: C24H34N4O8MOLECULAR WEIGHT:506.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [160806-26-8], RESEARCH AREA: ApoptosisREFERENCES: M.D.Mullican et al., Bioorg. Med. Chem. Letters, 4, 2359 (1994)