Learn More
Medchemexpress LLC Ac-[Nle}-Asp-His-{d-Phe}-Arg-Trp-Lys (lactam bridge: Asp4-Lys7) | 1607799-13-2 | 99.9% | 1085.22 | 100 MG
SDP
Catalog No. 5000388135
Encompass_Preferred
Bremelanotide Acetate is a melanocortin receptor agonist that can activate MC4R and increase dopamine release. It induces appetitive sexual behaviors, female mounting behavior, and repetitive self-grooming. This product can be used for the research of hypoactive sexual desire disorders.
  • Activates melanocortin receptors.
  • Increases dopamine release.
  • Induces appetitive sexual behaviors.
  • Induces female mounting behavior.
  • Induces repetitive self-grooming.
  • Can be used for the research of hypoactive sexual desire disorders.
  • Binds to MC3R and MC4R in a concentration-dependent manner.
  • Induces cAMP accumulation in cell-free assays.
Catalog No. 50-003-88135 Supplier Medchemexpress LLC Supplier No. HY18678A100MG
Bremelanotide Acetate is a melanocortin receptor agonist that can activate MC4R and increase dopamine release. It induces appetitive sexual behaviors, female mounting behavior, and repetitive self-grooming. This product can be used for the research of hypoactive sexual desire disorders.
  • Activates melanocortin receptors.
  • Increases dopamine release.
  • Induces appetitive sexual behaviors.
  • Induces female mounting behavior.
  • Induces repetitive self-grooming.
  • Can be used for the research of hypoactive sexual desire disorders.
  • Binds to MC3R and MC4R in a concentration-dependent manner.
  • Induces cAMP accumulation in cell-free assays.