Learn More
Enzo Life Sciences Ac-Nle-Pro-Nle-Asp-AMC (5mg)
SDP
Supplier:  Enzo Life Sciences BMLAW95550005
Encompass_Preferred
Highly specific substrate for the caspase-like peptidic activity of the 20S proteasome. Alternative name: Ac-nLPnLD-AMC. Purity: ≥97% (HPLC). Long Term Storage: -20°C.
Catalog No. 50-201-0333