Learn More
Bachem Ac-Nle-Pro-Nle-Asp-AMC
SDP
Supplier:  Bachem 4030084.0025
Encompass_Preferred
Ac-Nle-Pro-Nle-Asp-AMC, 355140-49-7, C33H45N5O9, Mr 655.75, 25mg. Ac-nLPnLD-AMC, substrate for proteasome.
Catalog No. 50-194-7060