Learn More
Medchemexpress LLC Ac-PAL-AMC | 1431362-79-6 | 1MG
SDP
Catalog No. 5000238712
Encompass_Preferred
Ac-PAL-AMC is a fluorogenic substrate specific for 20S proteasome LMP2/ 1i activity[1]
Catalog No. 50-002-38712 Supplier Medchemexpress LLC Supplier No. HY1230521MG
Ac-PAL-AMC is a fluorogenic substrate specific for 20S proteasome LMP2/ 1i activity[1]