Learn More
Medchemexpress LLC Ac-PAL-AMC | 1431362-79-6 | 99.9% | 498.57 | 5 MG
SDP
Catalog No. 5000364624
Encompass_Preferred
Ac-PAL-AMC is a fluorogenic substrate specific for 20S proteasome LMP2/β1i activity. It is intended for research use only.
  • Fluorogenic substrate
  • Specific for 20S proteasome LMP2/β1i activity
  • For research use only
Catalog No. 50-003-64624 Supplier Medchemexpress LLC Supplier No. 5MGHY123052
Ac-PAL-AMC is a fluorogenic substrate specific for 20S proteasome LMP2/β1i activity. It is intended for research use only.
  • Fluorogenic substrate
  • Specific for 20S proteasome LMP2/β1i activity
  • For research use only