Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-Phe-Lys-OH | 14287-21-9 | CB9470929 | 99.74% | 335.40 | 100 MG
SDP

Catalog No. 5000398838
Encompass_Preferred

Ac-Phe-Lys-OH is an inverted Aspartame type sweetener, provided as a white to off-white solid.

  • High purity of 99.74% (LCMS)
  • Molecular formula: C17H25N3O4
  • Molecular weight: 335.40
  • CAS number: 14287-21-9
  • Store powder at -80°C for 2 years or -20°C for 1 year
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-98838 Supplier Medchemexpress LLC Supplier No. HYP4840100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ac-Phe-Lys-OH is an inverted Aspartame type sweetener, provided as a white to off-white solid.

  • High purity of 99.74% (LCMS)
  • Molecular formula: C17H25N3O4
  • Molecular weight: 335.40
  • CAS number: 14287-21-9
  • Store powder at -80°C for 2 years or -20°C for 1 year
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.