Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-Phe-Lys-Pro-Leu-Ala-Ala-Ala-Arg | 2891606-02-1 | MFCD34676951 | 99.8% | 915.09 g/mol | C43H70N12O10 | 5 MG
SDP

Catalog No. 5000221169
Encompass_Preferred

BM213 is an acetylated octapeptide agonist of the complement C5a receptor 1 (C5aR1) employed in research to stimulate neutrophil activation and promote neutrophil extracellular trap (NET) formation. It is provided as a high-purity solid with characterized molecular properties and solubility suitable for in vitro and in vivo studies.

  • Peptide sequence: Ac-Phe-Lys-Pro-Leu-Ala-Ala-Ala-Arg.
  • High purity (99.8%).
  • Molecular weight 915.09 g/mol.
  • CAS number 2891606-02-1.
  • Soluble in water and PBS; may require sonication.
  • Storage: solid at -20°C; solutions stored at -80°C for long-term stability.
  • For research use only; not for human or diagnostic use.

Catalog No. 50-002-21169 Supplier Medchemexpress LLC Supplier No. HY1452375MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BM213 is an acetylated octapeptide agonist of the complement C5a receptor 1 (C5aR1) employed in research to stimulate neutrophil activation and promote neutrophil extracellular trap (NET) formation. It is provided as a high-purity solid with characterized molecular properties and solubility suitable for in vitro and in vivo studies.

  • Peptide sequence: Ac-Phe-Lys-Pro-Leu-Ala-Ala-Ala-Arg.
  • High purity (99.8%).
  • Molecular weight 915.09 g/mol.
  • CAS number 2891606-02-1.
  • Soluble in water and PBS; may require sonication.
  • Storage: solid at -20°C; solutions stored at -80°C for long-term stability.
  • For research use only; not for human or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.