Learn More
Carbosynth LLC. Ac-Phe-Thiaphe-OH, 25mg, MW: (M.W. 372.45), Molecular formula: C19H20N2O4S, CAS# [108906-59-8]
SDP
Supplier:  Carbosynth LLC. STP3621PI25MG
Encompass_Preferred
Synonyms: N-Acetyl-L-Phenylalanyl-L-3-Thiaphenylalanine, Application: Dipeptide Mimetic Substrate for Carboxypeptidase A, Storage Temperature: -20°C, References: P. Shamamian, S. Marcus, E. Deutsch, T. Maldonado, A. Liu, J. Stewart, K. Eng, and C. Gilvarg, Presented at the Annual Meeting of the Society for Surgery of the Alimentary Tract, May 1998., K.S. Brown, W.D. Kingsbury, N.M. Hall, G.L. Dunn, and C. Gilvarg, Anal. Biochem., 161, 219 (1987)., S.Y. Hwang, W.D. Kingsbury, N.M. Hall, D.R. Jakas, G.L. Dunn, and C. Gilvarg, Anal. Biochem., 154, 552 (1986).
Catalog No. 50-168-7165