Learn More
CPC Scientific Ac-Pro-Leu-Gly-((S)-2-mercapto-4-methyl-pentanoyl)-Leu-Gly-OEt 0.5MG
SDP
Catalog No. 502110522
Encompass
SEQUENCE: Ac-Pro-Leu-Gly-((S)-2-mercapto-4-methyl-pentanoyl)-Leu-Gly-OEtONE-LETTER SEQUENCE: Ac-PLG-((S)-2-mercapto-4-methyl-pentanoyl)-LG-OEtMOLECULAR FORMULA: C31H53N5O8S1MOLECULAR WEIGHT: 655.85STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [98992-65-5]SYNONYMS: RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES:SKU(s): SUBS-041A, SUBS-041B
Catalog No. 50-211-0522 Supplier CPC Scientific Supplier No. SUBS041A
Add to Cart
Add to Cart
SEQUENCE: Ac-Pro-Leu-Gly-((S)-2-mercapto-4-methyl-pentanoyl)-Leu-Gly-OEtONE-LETTER SEQUENCE: Ac-PLG-((S)-2-mercapto-4-methyl-pentanoyl)-LG-OEtMOLECULAR FORMULA: C31H53N5O8S1MOLECULAR WEIGHT: 655.85STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [98992-65-5]SYNONYMS: RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES:SKU(s): SUBS-041A, SUBS-041B