Learn More
AdipoGen Ac-Pro-Ala-Leu-AMC (5 mg)
SDP
Supplier:  AdipoGen AGCP30036M005
Encompass_Preferred
Chemical. CAS: 1431362-79-6. Formula: C26H34N4O6. MW: 498.6. Ac-PAL-AMC is a fluorogenic peptide substrate for measuring caspase-like activity of the immunoproteasome. Hydrolysis of this substrate by the beta1i/LMP2-subunit of the immunoproteasome is monitored by observing fluorescence at an Excitation wavelength of 345nm and Emission at 445nm. This substrate is specific to the immunoproteasome and is not hydrolyzed efficiently by the constitutive proteasome.
Catalog No. 50-205-3732