Learn More
Cayman Chemical Ac-QPKK(Ac)-AMC (trifluoroacetate salt), Purity: ≥95%, 5 mg
SDP
Catalog No. 502990497
Encompass_Preferred
A fluorogenic substrate for SIRT1, SIRT2, and SIRT3; AMC is released upon enzymatic cleavage by SIRT1, SIRT2, or SIRT3 and its fluorescence can be used to quantify SIRT1, SIRT2, and SIRT3 activity; ex/em = 340-360/440-460 nm, respectively
Catalog No. 50-299-0497 Supplier Cayman Chemical Supplier No. 307905 MG
A fluorogenic substrate for SIRT1, SIRT2, and SIRT3; AMC is released upon enzymatic cleavage by SIRT1, SIRT2, or SIRT3 and its fluorescence can be used to quantify SIRT1, SIRT2, and SIRT3 activity; ex/em = 340-360/440-460 nm, respectively